Learn More
Trajan Scientific Smart SPME Arrow 1.10 Grey 1 Pc
Catalog No. 501920797
Encompass
Smart SPME Arrow 1.10mm: Acrylate (Polyacrylate) Phase thickness 100µm Phase length 20mm color code: grey pack of 1 pc
Catalog No. 501920797 Supplier Trajan Scientific Supplier No. SARR11A100/20P1
Smart SPME Arrow 1.10mm: Acrylate (Polyacrylate) Phase thickness 100µm Phase length 20mm color code: grey pack of 1 pc