Learn More
Trajan Scientific Smart SPME Arrow 1.10 Grey 5 Pcs
Catalog No. 501920799
Encompass
Smart SPME Arrow 1.10mm: Acrylate (Polyacrylate) Phase thickness 100µm Phase length 20mm color code: grey pack of 5 pcs
Catalog No. 501920799 Supplier Trajan Scientific Supplier No. SARR11A100/20P5
Smart SPME Arrow 1.10mm: Acrylate (Polyacrylate) Phase thickness 100µm Phase length 20mm color code: grey pack of 5 pcs