Learn More
Trajan Scientific Smart SPME Arrow 1.10 Lt Bl 1 Pc
Catalog No. 501920800
Encompass
Smart SPME Arrow 1.10mm: Carbon WR/PDMS (Carbon Wide Range) Phase thickness 120µm Phase length 20mm color code: light blue pack of 1 pc
Catalog No. 501920800 Supplier Trajan Scientific Supplier No. SARR11CWR120/20P1
Smart SPME Arrow 1.10mm: Carbon WR/PDMS (Carbon Wide Range) Phase thickness 120µm Phase length 20mm color code: light blue pack of 1 pc