Learn More
Trajan Scientific Smart SPME Arrow 1.10 Red 1 Pc
Catalog No. 501920809
Encompass
Smart SPME Arrow 1.10mm: PDMS (Polydimethylsiloxane) Phase thickness 100µm Phase length 20mm color code: red pack of 1 pc
Catalog No. 501920809 Supplier Trajan Scientific Supplier No. SARR11P100/20P1
Smart SPME Arrow 1.10mm: PDMS (Polydimethylsiloxane) Phase thickness 100µm Phase length 20mm color code: red pack of 1 pc