Learn More
Trajan Scientific Smart SPME Arrow 1.10 Red 3 Pcs
Catalog No. 501920810
Encompass
Smart SPME Arrow 1.10mm: PDMS (Polydimethylsiloxane) Phase thickness 100µm Phase length 20mm color code: red pack of 3 pcs
Catalog No. 501920810 Supplier Trajan Scientific Supplier No. SARR11P100/20P3
Smart SPME Arrow 1.10mm: PDMS (Polydimethylsiloxane) Phase thickness 100µm Phase length 20mm color code: red pack of 3 pcs