Learn More
Trajan Scientific Smart SPME Arrow 1.10 Red 5 Pcs
Catalog No. 501920811
Encompass
Smart SPME Arrow 1.10mm: PDMS (Polydimethylsiloxane) Phase thickness 100µm Phase length 20mm color code: red pack of 5 pcs
Catalog No. 501920811 Supplier Trajan Scientific Supplier No. SARR11P100/20P5
Smart SPME Arrow 1.10mm: PDMS (Polydimethylsiloxane) Phase thickness 100µm Phase length 20mm color code: red pack of 5 pcs