Learn More
Trajan Scientific Smart SPME Arrow 1.10 Vlt 1 Pc
Catalog No. 501920803
Encompass
Smart SPME Arrow 1.10mm: DVB/PDMS (Divinylbenzene) Phase thickness 120µm Phase length 20mm color code: violett pack of 1 pc
Catalog No. 501920803 Supplier Trajan Scientific Supplier No. SARR11DVB120/20P1
Smart SPME Arrow 1.10mm: DVB/PDMS (Divinylbenzene) Phase thickness 120µm Phase length 20mm color code: violett pack of 1 pc