Learn More
Trajan Scientific Smart SPME Arrow 1.10 Vlt 3 Pcs
Catalog No. 501920804
Encompass
Smart SPME Arrow 1.10mm: DVB/PDMS (Divinylbenzene) Phase thickness 120µm Phase length 20mm color code: violett pack of 3 pcs
Catalog No. 501920804 Supplier Trajan Scientific Supplier No. SARR11DVB120/20P3
Smart SPME Arrow 1.10mm: DVB/PDMS (Divinylbenzene) Phase thickness 120µm Phase length 20mm color code: violett pack of 3 pcs