Learn More
Trajan Scientific Smart SPME Arrow 1.10 Vlt 5 Pcs
Catalog No. 501920805
Encompass
Smart SPME Arrow 1.10mm: DVB/PDMS (Divinylbenzene) Phase thickness 120µm Phase length 20mm color code: violett pack of 5 pcs
Catalog No. 501920805 Supplier Trajan Scientific Supplier No. SARR11DVB120/20P5
Smart SPME Arrow 1.10mm: DVB/PDMS (Divinylbenzene) Phase thickness 120µm Phase length 20mm color code: violett pack of 5 pcs