Learn More
Trajan Scientific Smart SPME Arrow 1.50 Dk Gry 3Pk
Catalog No. 501920821
Encompass
Smart SPME Arrow 1.50mm wide sleeve: DVB/C-WR/PDMS (Divinylbenzene/Carbon Wide Range/Polydimethylsiloxane) Phase thickness 120µm Phase length 20mm color code: dark grey pack of 3 pcs
Catalog No. 501920821 Supplier Trajan Scientific Supplier No. SARR15DVB/CWR120/20P3
Smart SPME Arrow 1.50mm wide sleeve: DVB/C-WR/PDMS (Divinylbenzene/Carbon Wide Range/Polydimethylsiloxane) Phase thickness 120µm Phase length 20mm color code: dark grey pack of 3 pcs