Learn More
Trajan Scientific Smart SPME Arrow 1.50 Lt Bl 1 Pc
Catalog No. 501920814
Encompass
Smart SPME Arrow 1.50mm wide sleeve: Carbon WR/PDMS (Carbon Wide Range) Phase thickness 120µm Phase length 20mm color code: light blue pack of 1 pc
Catalog No. 501920814 Supplier Trajan Scientific Supplier No. SARR15CWR120/20P1
Smart SPME Arrow 1.50mm wide sleeve: Carbon WR/PDMS (Carbon Wide Range) Phase thickness 120µm Phase length 20mm color code: light blue pack of 1 pc