Learn More
Trajan Scientific Smart SPME Arrow 1.50 Lt Bl 5Pk
Catalog No. 501920816
Encompass
Smart SPME Arrow 1.50mm wide sleeve: Carbon WR/PDMS (Carbon Wide Range) Phase thickness 120µm Phase length 20mm color code: light blue pack of 5 pcs
Catalog No. 501920816 Supplier Trajan Scientific Supplier No. SARR15CWR120/20P5
Smart SPME Arrow 1.50mm wide sleeve: Carbon WR/PDMS (Carbon Wide Range) Phase thickness 120µm Phase length 20mm color code: light blue pack of 5 pcs