Learn More
Trajan Scientific Smart SPME Arrow 1.50 Red 1 Pc
Catalog No. 501920823
Encompass
Smart SPME Arrow 1.50mm wide sleeve: PDMS (Polydimethylsiloxane) Phase thickness 100µm Phase length 20mm color code: red pack of 1 pc
Catalog No. 501920823 Supplier Trajan Scientific Supplier No. SARR15P100/20P1
Smart SPME Arrow 1.50mm wide sleeve: PDMS (Polydimethylsiloxane) Phase thickness 100µm Phase length 20mm color code: red pack of 1 pc