Learn More
Trajan Scientific Smart SPME Arrow 1.50 Red 3 Pcs
Catalog No. 501920824
Encompass
Smart SPME Arrow 1.50mm wide sleeve: PDMS (Polydimethylsiloxane) Phase thickness 100µm Phase length 20mm color code: red pack of 3 pcs
Catalog No. 501920824 Supplier Trajan Scientific Supplier No. SARR15P100/20P3
Smart SPME Arrow 1.50mm wide sleeve: PDMS (Polydimethylsiloxane) Phase thickness 100µm Phase length 20mm color code: red pack of 3 pcs