Learn More
Trajan Scientific Smart SPME Arrow 1.50 Red 5 Pcs
Catalog No. 501920825
Encompass
Smart SPME Arrow 1.50mm wide sleeve: PDMS (Polydimethylsiloxane) Phase thickness 100µm Phase length 20mm color code: red pack of 5 pcs
Catalog No. 501920825 Supplier Trajan Scientific Supplier No. SARR15P100/20P5
Smart SPME Arrow 1.50mm wide sleeve: PDMS (Polydimethylsiloxane) Phase thickness 100µm Phase length 20mm color code: red pack of 5 pcs