Learn More
Trajan Scientific Smart SPME Arrow 1.50 Vlt 1 Pc
Catalog No. 501920817
Encompass
Smart SPME Arrow 1.50mm wide sleeve: DVB/PDMS (Divinylbenzene) Phase thickness 120µm Phase length 20mm color code: violett pack of 1 pc
Catalog No. 501920817 Supplier Trajan Scientific Supplier No. SARR15DVB120/20P1
Smart SPME Arrow 1.50mm wide sleeve: DVB/PDMS (Divinylbenzene) Phase thickness 120µm Phase length 20mm color code: violett pack of 1 pc