Learn More
Trajan Scientific Smart SPME Arrow 1.50 Vlt 3 Pcs
Catalog No. 501920818
Encompass
Smart SPME Arrow 1.50mm wide sleeve: DVB/PDMS (Divinylbenzene) Phase thickness 120µm Phase length 20mm color code: violett pack of 3 pcs
Catalog No. 501920818 Supplier Trajan Scientific Supplier No. SARR15DVB120/20P3
Smart SPME Arrow 1.50mm wide sleeve: DVB/PDMS (Divinylbenzene) Phase thickness 120µm Phase length 20mm color code: violett pack of 3 pcs