Learn More
Trajan Scientific Smart SPME Arrow 1.50 Vlt 5 Pcs
Catalog No. 501920819
Encompass
Smart SPME Arrow 1.50mm wide sleeve: DVB/PDMS (Divinylbenzene) Phase thickness 120µm Phase length 20mm color code: violett pack of 3 pcs
Catalog No. 501920819 Supplier Trajan Scientific Supplier No. SARR15DVB120/20P5
Smart SPME Arrow 1.50mm wide sleeve: DVB/PDMS (Divinylbenzene) Phase thickness 120µm Phase length 20mm color code: violett pack of 3 pcs