Learn More
Trajan Scientific Smart SPME Arrow Variety 5 Pack
Catalog No. 501920813
Encompass
Collection of five different Smart SPME Arrows with phase length of 20mm: 1 pc SPME Arrow 1.10mm Acrylate (Polyacrylate) Phase thickness 100µm 1 pc SPME Arrow 1.10mm Carbon WR/PDMS (Carbon Wide Range / Polydimethylsiloxane) Phase thickness 120µm 1 pc SPME Arrow 1.10mm DVB/PDMS (Divinylbenzene / Polydimethylsiloxan) Phase thickness 120µm 1 pc SPME Arrow 1.10mm PDMS (Polydimethylsiloxane) Phase thickness 100µm 1 pc SPME Arrow 1.10mm DVB/Carbon WR/PDMS (Divinylbenzene/ Carbon wide range Polydimethylsiloxane) Phase thickness 120µm
Catalog No. 501920813 Supplier Trajan Scientific Supplier No. SARR1115SEL5S2
Collection of five different Smart SPME Arrows with phase length of 20mm: 1 pc SPME Arrow 1.10mm Acrylate (Polyacrylate) Phase thickness 100µm 1 pc SPME Arrow 1.10mm Carbon WR/PDMS (Carbon Wide Range / Polydimethylsiloxane) Phase thickness 120µm 1 pc SPME Arrow 1.10mm DVB/PDMS (Divinylbenzene / Polydimethylsiloxan) Phase thickness 120µm 1 pc SPME Arrow 1.10mm PDMS (Polydimethylsiloxane) Phase thickness 100µm 1 pc SPME Arrow 1.10mm DVB/Carbon WR/PDMS (Divinylbenzene/ Carbon wide range Polydimethylsiloxane) Phase thickness 120µm