Learn More
Trajan Scientific Smart SPME Fib 100Um Red 1 Pc
Catalog No. 501920870
Encompass
Smart SPME fiber PDMS (Polydimethylsiloxane) Fiber thickness 100µm Fiber length 10mm color code: red pack of 1pc
Catalog No. 501920870 Supplier Trajan Scientific Supplier No. SFIBP100/10P1
Smart SPME fiber PDMS (Polydimethylsiloxane) Fiber thickness 100µm Fiber length 10mm color code: red pack of 1pc