Learn More
Trajan Scientific Smart SPME Fib 100Um Red 3 Pcs
Catalog No. 501920871
Encompass
Smart SPME fiber PDMS (Polydimethylsiloxane) Fiber thickness 100µm Fiber length 10mm color code: red pack of 3pcs
Catalog No. 501920871 Supplier Trajan Scientific Supplier No. SFIBP100/10P3
Smart SPME fiber PDMS (Polydimethylsiloxane) Fiber thickness 100µm Fiber length 10mm color code: red pack of 3pcs