Learn More
Trajan Scientific Smart SPME Fib 100Um Red 5 Pcs
Catalog No. 501920872
Encompass
Smart SPME fiber PDMS (Polydimethylsiloxane) Fiber thickness 100µm Fiber length 10mm color code: red pack of 5pcs
Catalog No. 501920872 Supplier Trajan Scientific Supplier No. SFIBP100/10P5
Smart SPME fiber PDMS (Polydimethylsiloxane) Fiber thickness 100µm Fiber length 10mm color code: red pack of 5pcs