Learn More
Trajan Scientific Smart SPME Fib 30Um Gold 1 Pc
Catalog No. 501920873
Encompass
Smart SPME fiber PDMS (Polydimethylsiloxane) Fiber thickness 30µm Fiber length 10mm color code: golden pack of 1pc
Catalog No. 501920873 Supplier Trajan Scientific Supplier No. SFIBP30/10P1
Smart SPME fiber PDMS (Polydimethylsiloxane) Fiber thickness 30µm Fiber length 10mm color code: golden pack of 1pc