Learn More
Trajan Scientific Smart SPME Fib 30Um Gold 3 Pcs
Catalog No. 501920874
Encompass
Smart SPME fiber PDMS (Polydimethylsiloxane) Fiber thickness 30µm Fiber length 10mm color code: golden pack of 3pcs
Catalog No. 501920874 Supplier Trajan Scientific Supplier No. SFIBP30/10P3
Smart SPME fiber PDMS (Polydimethylsiloxane) Fiber thickness 30µm Fiber length 10mm color code: golden pack of 3pcs