Learn More
Trajan Scientific Smart SPME Fib 30Um Gold 5 Pcs
Catalog No. 501920875
Encompass
Smart SPME fiber PDMS (Polydimethylsiloxane) Fiber thickness 30µm Fiber length 10mm color code: golden pack of 5pcs
Catalog No. 501920875 Supplier Trajan Scientific Supplier No. SFIBP30/10P5
Smart SPME fiber PDMS (Polydimethylsiloxane) Fiber thickness 30µm Fiber length 10mm color code: golden pack of 5pcs