Learn More
Trajan Scientific Smart SPME Fib 7Um Grn 1 Pc
Catalog No. 501920876
Encompass
Smart SPME fiber PDMS (Polydimethylsiloxane ) Fiber thickness 7µm Fiber length 10mm color code: green pack of 1pc
Catalog No. 501920876 Supplier Trajan Scientific Supplier No. SFIBP7/10P1
Smart SPME fiber PDMS (Polydimethylsiloxane ) Fiber thickness 7µm Fiber length 10mm color code: green pack of 1pc