Learn More
Trajan Scientific Smart SPME Fib 7Um Grn 3 Pcs
Catalog No. 501920877
Encompass
Smart SPME fiber PDMS (Polydimethylsiloxane) Fiber thickness 7µm Fiber length 10mm color code: green pack of 3pcs
Catalog No. 501920877 Supplier Trajan Scientific Supplier No. SFIBP7/10P3
Smart SPME fiber PDMS (Polydimethylsiloxane) Fiber thickness 7µm Fiber length 10mm color code: green pack of 3pcs