Learn More
Trajan Scientific Smart SPME Fib 7Um Grn 5 Pcs
Catalog No. 501920878
Encompass
Smart SPME fiber PDMS (Polydimethylsiloxane) Fiber thickness 7µm Fiber length 10mm color code: green pack of 5pcs
Catalog No. 501920878 Supplier Trajan Scientific Supplier No. SFIBP7/10P5
Smart SPME fiber PDMS (Polydimethylsiloxane) Fiber thickness 7µm Fiber length 10mm color code: green pack of 5pcs