Learn More
Trajan Scientific Smart SPME Fib 85Um Grey 1 Pc
Catalog No. 501920858
Encompass
Smart SPME fiberAcrylate (Polyacrylate) Fiber thickness 85µm Fiber length 10mm color code: grey pack of 1pc
Catalog No. 501920858 Supplier Trajan Scientific Supplier No. SFIBA85/10P1
Smart SPME fiberAcrylate (Polyacrylate) Fiber thickness 85µm Fiber length 10mm color code: grey pack of 1pc