Learn More
Trajan Scientific Smart SPME Fib 85Um Grey 3 Pcs
Catalog No. 501920859
Encompass
Smart SPME fiber Acrylate (Polyacrylate) Fiber thickness 85µm Fiber length 10mm color code: grey pack of 3pcs
Catalog No. 501920859 Supplier Trajan Scientific Supplier No. SFIBA85/10P3
Smart SPME fiber Acrylate (Polyacrylate) Fiber thickness 85µm Fiber length 10mm color code: grey pack of 3pcs