Learn More
Trajan Scientific Smart SPME Fib Dvb/C-Wr/Pdms 5
Catalog No. 501920869
Encompass
Smart SPME fiber DVB/C-WR/PDMS (Divinylbenzene/Carbon Wide Range/Polydimethylsiloxane) Fiber thickness 50/30µm Fiber length 10mm color code: dark gray pack of 5pcs
Catalog No. 501920869 Supplier Trajan Scientific Supplier No. SFIBDVB/CWR80/10P5
Smart SPME fiber DVB/C-WR/PDMS (Divinylbenzene/Carbon Wide Range/Polydimethylsiloxane) Fiber thickness 50/30µm Fiber length 10mm color code: dark gray pack of 5pcs