Learn More
Trajan Scientific Smart SPME Fib Dvb/Pdms Vlt 1
Catalog No. 501920864
Encompass
Smart SPME fiber DVB/PDMS (Divinylbenzene / Polydimethylsiloxane) Fiber thickness 65µm Fiber length 10mm color code: violett pack of 1pc
Catalog No. 501920864 Supplier Trajan Scientific Supplier No. SFIBDVB65/10P1
Smart SPME fiber DVB/PDMS (Divinylbenzene / Polydimethylsiloxane) Fiber thickness 65µm Fiber length 10mm color code: violett pack of 1pc