Learn More
Trajan Scientific Smart SPME Fib Dvb/Pdms Vlt 3
Catalog No. 501920865
Encompass
Smart SPME fiber DVB/PDMS (Divinylbenzene / Polydimethylsiloxane) Fiber thickness 65µm Fiber length 10mm color code: violett pack of 3pcs
Catalog No. 501920865 Supplier Trajan Scientific Supplier No. SFIBDVB65/10P3
Smart SPME fiber DVB/PDMS (Divinylbenzene / Polydimethylsiloxane) Fiber thickness 65µm Fiber length 10mm color code: violett pack of 3pcs