Learn More
Trajan Scientific Smart SPME Fib Dvb/Pdms Vlt 5
Catalog No. 501920866
Encompass
Smart SPME fiber DVB/PDMS (Divinylbenzene / Polydimethylsiloxane) Fiber thickness 65µm Fiber length 10mm color code: violett pack of 5pcs
Catalog No. 501920866 Supplier Trajan Scientific Supplier No. SFIBDVB65/10P5
Smart SPME fiber DVB/PDMS (Divinylbenzene / Polydimethylsiloxane) Fiber thickness 65µm Fiber length 10mm color code: violett pack of 5pcs