Learn More
Trajan Scientific Smart SPME Fib Variety 5 Pack
Catalog No. 501920880
Encompass
Collection of five different Smart SPME fibers with fiber length of 10mm and fiber thickness of: PDMS: 100µm DVB: 65µm Carbon WR: 95µm Acrylate: 85µm DVB/Carbon WR:80µm
Catalog No. 501920880 Supplier Trajan Scientific Supplier No. SFIBSEL5S2
Collection of five different Smart SPME fibers with fiber length of 10mm and fiber thickness of: PDMS: 100µm DVB: 65µm Carbon WR: 95µm Acrylate: 85µm DVB/Carbon WR:80µm