Learn More
ZORO TOOLS General Purpose Lubricant, 12 oz, Aerosol Spray Can, Mineral Oil Base, 300 Degrees F Max Op Temp, Amber, Original Formula
Catalog No. NC2271328
Encompass
General Purpose Lubricant, 12 oz, Aerosol Spray Can, Mineral Oil Base, 300 Degrees F Max Op Temp, Amber, Original Formula
This item is not returnable. View return policy
Catalog No. NC2271328 Supplier ZORO TOOLS Supplier No. G5417596
General Purpose Lubricant, 12 oz, Aerosol Spray Can, Mineral Oil Base, 300 Degrees F Max Op Temp, Amber, Original Formula