Learn More
Trajan Scientific Smart Syringe 1.2Ul 23S
Catalog No. 501920837
Encompass
1.2µL Smart Syringe with fixed needle for Tool D7/85: Needle length 85mm gauge 26P scale length 36mm Point Style conical Replacement for PAL3-SYH-207888
Catalog No. 501920837 Supplier Trajan Scientific Supplier No. SF185B26PCO
1.2µL Smart Syringe with fixed needle for Tool D7/85: Needle length 85mm gauge 26P scale length 36mm Point Style conical Replacement for PAL3-SYH-207888