Learn More
Trajan Scientific Smart Syringe 1000Ul 23G
Catalog No. 501920848
Encompass
1000uL Smart Syringe with fixed needle for Tool D8/57: Needle length 57mm PTFE plunger gauge 23 scale length 60mm Point Style conical (Instrument: Pal3)
Catalog No. 501920848 Supplier Trajan Scientific Supplier No. SF100057T23CO
1000uL Smart Syringe with fixed needle for Tool D8/57: Needle length 57mm PTFE plunger gauge 23 scale length 60mm Point Style conical (Instrument: Pal3)