Learn More
Trajan Scientific Smart Syringe 100Ul 26S 57
Catalog No. 501920845
Encompass
100uL Smart Syringe with fixed needle for Tool D7/57: Needle length 57mm PTFE plunger gauge 26S scale length 60mm Point Style conical (Instrument: Pal3)
Catalog No. 501920845 Supplier Trajan Scientific Supplier No. SF10057T26SCO
100uL Smart Syringe with fixed needle for Tool D7/57: Needle length 57mm PTFE plunger gauge 26S scale length 60mm Point Style conical (Instrument: Pal3)