Learn More
Trajan Scientific Smart Syringe 100Ul 26S 85
Catalog No. 501920846
Encompass
100uL Smart Syringe with fixed needle for Tool D7/85: Needle length 85mm PTFE plunger gauge 26S scale length 60mm Point Style conical (Instrument: Pal3)
Catalog No. 501920846 Supplier Trajan Scientific Supplier No. SF10085T26SCO
100uL Smart Syringe with fixed needle for Tool D7/85: Needle length 85mm PTFE plunger gauge 26S scale length 60mm Point Style conical (Instrument: Pal3)