Learn More
Trajan Scientific Smart Syringe 10Ul 22S
Catalog No. 501920840
Encompass
10uL Smart Syringe with fixed needle for Tool D7/57: Needle length 57mm PTFE plunger gauge 22S scale length 60mm Point Style flat (Instrument: Pal3)
Catalog No. 501920840 Supplier Trajan Scientific Supplier No. SF1057T22SFL
10uL Smart Syringe with fixed needle for Tool D7/57: Needle length 57mm PTFE plunger gauge 22S scale length 60mm Point Style flat (Instrument: Pal3)