Learn More
Trajan Scientific Smart Syringe 10Ul 26S 85 Ptfe
Catalog No. 501920844
Encompass
10uL Smart Syringe with fixed needle for Tool D7/85: Needle length 85mm PTFE plunger gauge 26S scale length 60mm Point Style conical (Instrument: Pal3)
Catalog No. 501920844 Supplier Trajan Scientific Supplier No. SF1085T26SCO
10uL Smart Syringe with fixed needle for Tool D7/85: Needle length 85mm PTFE plunger gauge 26S scale length 60mm Point Style conical (Instrument: Pal3)