Learn More
Trajan Scientific Smart Syringe 10Ul 26S 85
Catalog No. 501920843
Encompass
10uL Smart Syringe with fixed needle for Tool D7/85: Needle length 85mm metal plunger gauge 26S scale length 54mm Point Style conical (Instrument: Pal3)
Catalog No. 501920843 Supplier Trajan Scientific Supplier No. SF1085M26SCO
10uL Smart Syringe with fixed needle for Tool D7/85: Needle length 85mm metal plunger gauge 26S scale length 54mm Point Style conical (Instrument: Pal3)