Learn More
Trajan Scientific Smart Syringe 10Ul 26S Co
Catalog No. 501920842
Encompass
10uL Smart Syringe with fixed needle for Tool D7/57: Needle length 57mm PTFE plunger gauge 26S scale length 60mm Point Style conical (Instrument: Pal3)
Catalog No. 501920842 Supplier Trajan Scientific Supplier No. SF1057T26SCO
10uL Smart Syringe with fixed needle for Tool D7/57: Needle length 57mm PTFE plunger gauge 26S scale length 60mm Point Style conical (Instrument: Pal3)