Learn More
Trajan Scientific Smart Syringe 250Ul 22G
Catalog No. 501920851
Encompass
250uL Smart Syringe with fixed needle for Tool D8/57: Needle length 57mm PTFE plunger gauge 22 scale length 60mm Point Style flat (Instrument: Pal3)
Catalog No. 501920851 Supplier Trajan Scientific Supplier No. SF25057T22FL
250uL Smart Syringe with fixed needle for Tool D8/57: Needle length 57mm PTFE plunger gauge 22 scale length 60mm Point Style flat (Instrument: Pal3)