Learn More
Trajan Scientific Smart Syringe 250Ul 26G 57
Catalog No. 501920852
Encompass
250uL Smart Syringe with fixed needle for Tool D8/57: Needle length 57mm PTFE plunger gauge 26 scale length 60mm Point Style conical (Instrument: Pal3)
Catalog No. 501920852 Supplier Trajan Scientific Supplier No. SF25057T26CO
250uL Smart Syringe with fixed needle for Tool D8/57: Needle length 57mm PTFE plunger gauge 26 scale length 60mm Point Style conical (Instrument: Pal3)