Learn More
Trajan Scientific Smart Syringe 250Ul 26G 85
Catalog No. 501920853
Encompass
250uL Smart Syringe with fixed needle for Tool D8/85: Needle length 85mm PTFE plunger gauge 26 scale length 60mm Point Style conical (Instrument: Pal3)
Catalog No. 501920853 Supplier Trajan Scientific Supplier No. SF25085T26CO
250uL Smart Syringe with fixed needle for Tool D8/85: Needle length 85mm PTFE plunger gauge 26 scale length 60mm Point Style conical (Instrument: Pal3)