Learn More
Trajan Scientific Smart Syringe 5000Ul G22 Ptfe
Catalog No. 501920833
Encompass
5000uL Smart Syringe with fixed needle for Tool D18/57: Needle length 57mm PTFE plunger gauge 22 scale length 60mm Point Style flat (Instrument: Pal3)
Catalog No. 501920833 Supplier Trajan Scientific Supplier No. SC500057T22FL
5000uL Smart Syringe with fixed needle for Tool D18/57: Needle length 57mm PTFE plunger gauge 22 scale length 60mm Point Style flat (Instrument: Pal3)