Learn More
Medschenker™ Smart Transport Medium
SDP
Catalog No. 23000001
Change view
Click to view available options
Volume (Metric):
3 mL
10 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric)
23-000-001 3 mL
23-000-000 10 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 23-000-001 Supplier Medschenker™ Supplier No. STM30A
Add to Cart
Add to Cart
This item is not returnable. View return policy
For collection, transport and maintenance of viruses.
  • Medschenker™ STM includes antibiotics to help inhibit bacteria and fungal flora in the sample for the most accurate results
  • Medschenker™ STM is a stand-up and shatterproof tube enabling quality centrifugation of the sample
  • Patient sample material and particles are rapidly released and dispersed
  • It maintains organisms viability for 48 hours at room or refrigerated temperature
  • MedSchenker™ STM features a new cap-lock system for secure transport without the risk of leaking or spilling
Specifications
Sterility Sterile
Closure Type Screw Cap
Quantity 50/Pk., 1000/Cs.
Storage Requirements 2°C to 25°C
Volume (Metric) 3 mL
Material Plastic