Learn More
Electron Microscopy Sciences Smart Tweezers Quick-Test Auto-Scanning Tweezers
SDP
Supplier:  Electron Microscopy Sciences 7290701
Encompass_Preferred
Precise, non-magnetic tips reliably contact smallest SMD. The Quick Test tweezers gives users an easy way to sort and evaluate loose components and to perform on-board measurements and debugging. Surface mount devices are usually tiny and without wire leads, making it much more difficult to test and identify SMD than conventional components. Precise tips, made of non-magnetic steel, are able to pick and reliably contact even the smallest SMD components, or take measurements from the device already mounted on a board. The probe can also be used to test conventional components with wire leads too short to insert into the test terminals.
The tweezers measures AC/DC voltage, resistance, continunity & diode test and has auto scanning mode and auto range. Testing pins are gold plated to reduce resistance and prevent rust. Lithium battery CR2032 is included. The tweezers will automatically identify whether component is a resistor, capacitor or diode.
Catalog No. 50-240-45